4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I0856-100G I0856-25G
TCI0856-100GEA 429.76 CAD
TCI0856-100G TCI0856-25G
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride
Formula: C₃₁H₂₀O₈
MDL Number: MFCD00319167
CAS Number: 38103-06-9
EINECS: 253-781-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Methanolysis Method) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR