5-Methyl-3,4-diphenylisoxazole ≥98.0% (by GC)

Supplier: TCI America

M2830-1G M2830-5G
TCM2830-1GEA 52.14 CAD
TCM2830-1G TCM2830-5G
5-Methyl-3,4-diphenylisoxazole ≥98.0% (by GC)
5-Methyl-3,4-diphenylisoxazole
Formula: C₁₆H₁₃NO
MDL Number: MFCD18449672
CAS Number: 37928-17-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR