N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D4855-1G
D4855-200MG
TCD4855-1GEA
432.92
CAD
TCD4855-1G
TCD4855-200MG
N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine
Formula:
C₅₂H₃₆N₂ MW: 688.87 g/mol |
MDL Number:
MFCD16038221 CAS Number: 374592-88-8 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...