4-Fluorohippuric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Fluorobenzamido)acetic acid, N-(4-Fluorobenzoyl)glycine

F1162-1G F1162-200MG
TCF1162-1GEA 237 CAD
TCF1162-1G TCF1162-200MG
4-Fluorohippuric acid ≥98.0% (by GC, titration analysis)
4-Fluorohippuric acid
Formula: C₉H₈FNO₃
MW: 197.17 g/mol
MDL Number: MFCD00462263
CAS Number: 366-79-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR