4-Bromocarbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4752-1G
B4752-200MG
TCB4752-1GEA
157.05
CAD
TCB4752-1G
TCB4752-200MG
4-Bromocarbazole ≥98.0% (by GC)
4-Bromocarbazole
Formula:
C₁₂H₈BrN |
MDL Number:
MFCD18450175 CAS Number: 3652-89-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...