N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4237-1G B4237-200MG
TCB4237-1GEA 409.22 CAD
TCB4237-1G TCB4237-200MG
N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine ≥98.0% (by GC, titration analysis)
N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine
Formula: C₁₁H₁₅FN₂O₂
MDL Number: MFCD07787180
CAS Number: 362670-07-3

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR