2,2'-Bipyridine-3,3'-diol ≥98.0% (by GC)

Supplier: TCI America
Warning

B4255-250MG B4255-50MG
TCB4255-250MGEA 680.98 CAD
TCB4255-250MG TCB4255-50MG
2,2'-Bipyridine-3,3'-diol ≥98.0% (by GC)
2,2'-Bipyridine-3,3'-diol
Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00075177
CAS Number: 36145-03-6

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR