Tiaramide hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
T3176-100MG
T3176-25MG
TCT3176-100MGEA
586.18
CAD
TCT3176-100MG
TCT3176-25MG
Tiaramide hydrochloride ≥98.0% (by titrimetric analysis)
Tiaramide hydrochloride
Formula:
C₁₅H₁₈ClN₃O₃S·HCl Melting Pt: 160°C |
MDL Number:
MFCD01702832 CAS Number: 35941-71-0 EINECS: 252-802-7 Merck Index: 14,09424 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...