2,3,4,5-Tetramethoxytoluene ≥97.0% (by GC)
Supplier: TCI America
T2626-25G
T2626-5G
76220-522EA
124.82
CAD
76220-522
76220-524
2,3,4,5-Tetramethoxytoluene ≥97.0% (by GC)
2,3,4,5-Tetramethoxytoluene
Formula:
C₁₁H₁₆O₄ Boiling Pt: 120°C |
MDL Number:
MFCD07370118 CAS Number: 35896-58-3 |
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...