2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine ≥98.0% (by total nitrogen basis)

Supplier: TCI America

B4649-1G B4649-5G
TCB4649-1GEA 233.84 CAD
TCB4649-1G TCB4649-5G
2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine ≥98.0% (by total nitrogen basis)
2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine
Formula: C₃₆H₄₄N₂O₄
MDL Number: MFCD28386094
CAS Number: 358727-55-6

Order Now

Specification Test Results

Appearance Yellow to Amber powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR