O-(4-Nitrobenzoyl)hydroxylamine ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

N1013-1G N1013-5G
TCN1013-1GEA 77.42 CAD
TCN1013-1G TCN1013-5G
O-(4-Nitrobenzoyl)hydroxylamine ≥97.0% (by HPLC, total nitrogen)
O-(4-Nitrobenzoyl)hydroxylamine
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11976095
CAS Number: 35657-36-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR