2-Anilino-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2741-1G A2741-5G
TCA2741-1GEA 237.95 CAD
TCA2741-1G TCA2741-5G
2-Anilino-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
2-Anilino-9,9-dimethylfluorene
Formula: C₂₁H₁₉N
MDL Number: MFCD20040462
CAS Number: 355832-04-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR