6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D4980-1G D4980-200MG
TCD4980-1GEA 522.98 CAD
TCD4980-1G TCD4980-200MG
6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole ≥95.0% (by HPLC, total nitrogen)
6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole
Formula: C₃₆H₂₂Br₂N₂
MDL Number: MFCD30749258
CAS Number: 354135-75-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR