3,3-(Diphenyl)acrylonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

D4873-1G D4873-200MG
TCD4873-1GEA 357.08 CAD
TCD4873-1G TCD4873-200MG
3,3-(Diphenyl)acrylonitrile ≥98.0% (by GC)
3,3-(Diphenyl)acrylonitrile
Formula: C₁₅H₁₁N
Boiling Pt: 131°C
MDL Number: MFCD00045630
CAS Number: 3531-24-6

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR