N-(4-Fluorophenyl)glycine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F0960-1G F0960-5G
TCF0960-1GEA 217.95 CAD
TCF0960-1G TCF0960-5G
N-(4-Fluorophenyl)glycine ≥98.0% (by GC, titration analysis)
N-(4-Fluorophenyl)glycine
Formula: C₈H₈FNO₂
Melting Pt: 138°C
MDL Number: MFCD02255629
CAS Number: 351-95-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR