1,4-Bis(2-propynyloxy)benzene ≥98.0% (by GC)

Supplier: TCI America

B4607-1G B4607-5G
TCB4607-1GEA 160.37 CAD
TCB4607-1G TCB4607-5G
1,4-Bis(2-propynyloxy)benzene ≥98.0% (by GC)
1,4-Bis(2-propynyloxy)benzene
Formula: C₁₂H₁₀O₂
Boiling Pt: 202°C
Storage Temperature: Refrigerator
MDL Number: MFCD00224313
CAS Number: 34596-36-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR