2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5162-1G
D5162-5G
76219-200EA
91.64
CAD
76219-200
76219-202
2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole ≥98.0% (by GC)
2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole
Formula:
C₁₆H₁₇N |
CAS Number:
344928-74-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...