(S)-(-)-3-Amino-2-piperidone ≥98.0% (by GC)

Supplier: TCI America
Warning

A3171-1G A3171-5G
76219-316EA 399.74 CAD
76219-316 76219-318
(S)-(-)-3-Amino-2-piperidone ≥98.0% (by GC)
(S)-(-)-3-Amino-2-piperidone
Formula: C₅H₁₀N₂O
Melting Pt: 87°C
Storage Temperature: Freezer
MDL Number: MFCD09258624
CAS Number: 34294-79-6

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D -12.0 to -16.0 deg (C=0.5, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR