1-Iodopyrene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
I0977-1G
I0977-200MG
TCI0977-1GEA
557.11
CAD
TCI0977-1G
TCI0977-200MG
1-Iodopyrene ≥98.0% (by GC)
1-Iodopyrene
Formula:
C₁₆H₉I |
CAS Number:
34244-15-0 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...