9-Benzyl-3-bromo-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America

B5461-1G B5461-5G
TCB5461-1GEA 107.44 CAD
TCB5461-1G TCB5461-5G
9-Benzyl-3-bromo-9H-carbazole ≥98.0% (by GC)
9-Benzyl-3-bromo-9H-carbazole
Formula: C₁₉H₁₄BrN
MW: 336.23 g/mol
MDL Number: MFCD00821698
CAS Number: 339576-55-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR