4-Methoxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2828-1G M2828-5G
TCM2828-1GEA 91.64 CAD
TCM2828-1G TCM2828-5G
4-Methoxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-Methoxy-2-nitrobenzoic acid
Formula: C₈H₇NO₅
MDL Number: MFCD04972109
CAS Number: 33844-21-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR