(4-Hydroxyphenyl)phosphonic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H1559-1G H1559-200MG
76220-136EA 859.52 CAD
76220-136 76220-138
(4-Hydroxyphenyl)phosphonic acid ≥97.0% (by HPLC, titration analysis)
(4-Hydroxyphenyl)phosphonic acid
Formula: C₆H₇O₄P
MDL Number: MFCD01112120
CAS Number: 33795-18-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR