Sulbutiamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
S0964-25G
S0964-5G
TCS0964-25GEA
429.76
CAD
TCS0964-25G
TCS0964-5G
Sulbutiamine ≥98.0% (by HPLC, titration analysis)
Sulbutiamine
Formula:
C₃₂H₄₆N₈O₆S₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01726427 CAS Number: 3286-46-2 EINECS: 221-937-3 Merck Index: 14,09297 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...