1-Ethyl-4-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E1272-25G E1272-5G
76220-046EA 285.98 CAD
76220-046 76220-048
1-Ethyl-4-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Ethyl-4-methylpyridinium bromide
Formula: C₈H₁₂BrN
MW: 202.10 g/mol
MDL Number: MFCD18384825
CAS Number: 32353-49-4

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR