Triflusal ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Acetoxy-4-(trifluoromethyl)benzoic acid, Acetyl-4-(trifluoromethyl)salicylic acid, Drisgen
T3601-1G
T3601-250MG
TCT3601-1GEA
1011.2
CAD
TCT3601-1G
TCT3601-250MG
Triflusal ≥98.0% (by titrimetric analysis)
Triflusal
Formula:
C₁₀H₇F₃O₄ MW: 248.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00866793 CAS Number: 322-79-2 EINECS: 206-297-5 Merck Index: 14,09688 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...