Ethyl (Benzyloxy)acetate ≥98.0% (by GC)
Supplier: TCI America
E1225-1G
E1225-5G
TCE1225-1GEA
61.62
CAD
TCE1225-1G
TCE1225-5G
Ethyl (Benzyloxy)acetate ≥98.0% (by GC)
Ethyl (Benzyloxy)acetate
Formula:
C₁₁H₁₄O₃ Boiling Pt: 287°C Storage Temperature: Refrigerator |
MDL Number:
MFCD06204614 CAS Number: 32122-09-1 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Acid Value | max. 2.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...