1-(2-Nitrophenyl)ethanol ≥98.0% (by GC)

Supplier: TCI America

N1132-1G N1132-5G
76220-384EA 153.26 CAD
76220-384 76220-386
1-(2-Nitrophenyl)ethanol ≥98.0% (by GC)
1-(2-Nitrophenyl)ethanol
Formula: C₈H₉NO₃
Boiling Pt: 107°C
MDL Number: MFCD02683380
CAS Number: 3205-25-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR