2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine 98,0 HPLC, N

Supplier: TCI America

T3539-1G T3539-200MG
76220-674EA 478.74 CAD
76220-674 76220-676
2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine 98,0 HPLC, N
2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine
Formula: C₃₉H₂₇N₃
MW: 537.67 g/mol
CAS Number: 31274-51-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR