8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C3181-25G C3181-5G
TCC3181-25GEA 613.04 CAD
TCC3181-25G TCC3181-5G
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one ≥98.0% (by GC, titration analysis)
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
Formula: C₁₄H₁₀ClNO
Storage Temperature: Freezer
MDL Number: MFCD00800222
CAS Number: 31251-41-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR