Diphenyl Phenylphosphonate ≥98.0% (by GC)

Supplier: TCI America

D5454-1G D5454-200MG
TCD5454-1GEA 421.86 CAD
TCD5454-1G TCD5454-200MG
Diphenyl Phenylphosphonate ≥98.0% (by GC)
Diphenyl Phenylphosphonate
Formula: C₁₈H₁₅O₃P
Boiling Pt: 165°C
Melting Pt: 75°C
MDL Number: MFCD00216888
CAS Number: 3049-24-9
EINECS: 221-261-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR