2-Bromo-3-(2-ethylhexyl)thiophene ≥98.0% (by GC)

Supplier: TCI America

B4783-1G B4783-5G
TCB4783-1GEA 191.81 CAD
TCB4783-1G TCB4783-5G
2-Bromo-3-(2-ethylhexyl)thiophene ≥98.0% (by GC)
2-Bromo-3-(2-ethylhexyl)thiophene
Formula: C₁₂H₁₉BrS
Storage Temperature: Refrigerator
MDL Number: MFCD26743667
CAS Number: 303734-52-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR