Clinofibrate ≥98.0% (by HPLC)

Supplier: TCI America

C3100-1G C3100-200MG
TCC3100-1GEA 448.72 CAD
TCC3100-1G TCC3100-200MG
Clinofibrate ≥98.0% (by HPLC)
Clinofibrate
Formula: C₂₈H₃₆O₆
MDL Number: MFCD00865769
CAS Number: 30299-08-2
Merck Index: 14,02357

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR