17alpha-Hydroxyprogesterone Acetate ≥98.0% (by HPLC)

Supplier: TCI America

H1527-1G H1527-5G
TCH1527-1GEA 79 CAD
TCH1527-1G TCH1527-5G
17alpha-Hydroxyprogesterone Acetate ≥98.0% (by HPLC)
17alpha-Hydroxyprogesterone Acetate
Formula: C₂₃H₃₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00021152
CAS Number: 302-23-8
EINECS: 206-119-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 244.0 to 248.0 deg-C
Specific rotation [a]20/D +71.0 to +76.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR