Tianeptine sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 7-[[3-Chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl]amino]heptanoic acid S,S-dioxide sodium salt hydrate

T3131-1G T3131-200MG
TCT3131-1GEA 690.93 CAD
TCT3131-1G TCT3131-200MG
Tianeptine sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)
Tianeptine sodium salt hydrate
Formula: C₂₁H₂₄ClN₂NaO₄S·xH₂O
MW: 458.93 g/mol (anhy)
MDL Number: MFCD00879851
CAS Number: 30123-17-2
EINECS: 250-059-3
Merck Index: 14,09420

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Melting point 183.0 to 187.0 deg-C
Water 3.0 to 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR