2-Morpholinoethyl methacrylate ≥95.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Methacrylic acid 2-Morpholinoethyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M2901-1G M2901-5G
TCM2901-1GEA 94.8 CAD
TCM2901-1G TCM2901-5G
2-Morpholinoethyl methacrylate ≥95.0% (by GC) stabilised
2-Morpholinoethyl methacrylate
Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Boiling Pt: 81°C
MDL Number: MFCD00080577
CAS Number: 2997-88-8
EINECS: 221-069-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR