2-Isopropyl-2-methacryloyloxyadamantane ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Isopropyl-2-adamantyl methacrylate, Methacrylic acid 2-Isopropyl-2-adamantyl ester
I0962-25G
I0962-5G
TCI0962-25GEA
492.96
CAD
TCI0962-25G
TCI0962-5G
2-Isopropyl-2-methacryloyloxyadamantane ≥98.0% (by GC)
2-Isopropyl-2-methacryloyloxyadamantane
Formula:
C₁₇H₂₆O₂ MW: 262.39 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22571643 CAS Number: 297156-50-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...