1-(4-Fluorophenyl)-1,3-butanedione ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
(4-Fluorobenzoyl)acetone, 1-(4-Fluorophenyl)butane-1,3-dione
F0996-1G
F0996-5G
76220-074EA
282.82
CAD
76220-074
76220-076
1-(4-Fluorophenyl)-1,3-butanedione ≥97.0% (by GC)
1-(4-Fluorophenyl)-1,3-butanedione
Formula:
C₁₀H₉FO₂ Boiling Pt: 117°C |
MDL Number:
MFCD00463139 CAS Number: 29681-98-9 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...