4-(1H-Benzimidazol-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(1H-Benzo[d]imidazol-2-yl)aniline, 2-(4-Aminophenyl)benzimidazole
B5710-1G
B5710-5G
TCB5710-1GEA
268.6
CAD
TCB5710-1G
TCB5710-5G
4-(1H-Benzimidazol-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
4-(1H-Benzimidazol-2-yl)aniline
Formula:
C₁₃H₁₁N₃ MW: 209.25 g/mol |
MDL Number:
MFCD00454395 CAS Number: 2963-77-1 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 248.0 to 252.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...