4-Chloro-2,6-diphenylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3016-1G C3016-5G
TCC3016-1GEA 69.52 CAD
TCC3016-1G TCC3016-5G
4-Chloro-2,6-diphenylpyrimidine ≥98.0% (by GC)
4-Chloro-2,6-diphenylpyrimidine
Formula: C₁₆H₁₁ClN₂
MDL Number: MFCD00234890
CAS Number: 29509-91-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR