2-Chloro-4,6-diphenylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3123-1G C3123-5G
TCC3123-1GEA 67.94 CAD
TCC3123-1G TCC3123-5G
2-Chloro-4,6-diphenylpyrimidine ≥98.0% (by GC)
2-Chloro-4,6-diphenylpyrimidine
Formula: C₁₆H₁₁ClN₂
MDL Number: MFCD00030776
CAS Number: 2915-16-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR