Diethyl (2-Chlorobenzyl)phosphonate ≥98.0% (by GC)

Supplier: TCI America

D5265-1G D5265-5G
TCD5265-1GEA 127.98 CAD
TCD5265-1G TCD5265-5G
Diethyl (2-Chlorobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (2-Chlorobenzyl)phosphonate
Formula: C₁₁H₁₆ClO₃P
Boiling Pt: 146°C
MDL Number: MFCD22574983
CAS Number: 29074-98-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
IR to pass test
NMR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR