Ethyl 3-Oxo-3-(4-chlorophenyl)propionate 98,0 GC_ASSAY_METHOD

Supplier: TCI America

Synonyms: Ethyl 3-(4-chlorophenyl)-3-oxo-propionate, 3-Oxo-3-(4-chlorophenyl)propionic acid Ethyl ester

E1059-1G E1059-5G
TCE1059-1GEA 134.3 CAD
TCE1059-1G TCE1059-5G
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate 98,0 GC_ASSAY_METHOD
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate
Formula: C₁₁H₁₁ClO₃
MW: 226.66 g/mol
Boiling Pt: 123°C
Storage Temperature: Refrigerator
MDL Number: MFCD00018713
CAS Number: 2881-63-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR