4-Benzyl-2-hydroxymorpholin-3-one ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4918-1G
B4918-5G
TCB4918-1GEA
37.92
CAD
TCB4918-1G
TCB4918-5G
4-Benzyl-2-hydroxymorpholin-3-one ≥98.0% (by GC)
4-Benzyl-2-hydroxymorpholin-3-one
Formula:
C₁₁H₁₃NO₃ |
MDL Number:
MFCD07776435 CAS Number: 287930-73-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 134.0 to 138.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...