Tris(3,5-dimethyl-1-pyrazolyl)methane ≥98.0% (by GC)
Supplier: TCI America
T3025-1G
T3025-200MG
TCT3025-1GEA
331.09
CAD
TCT3025-1G
TCT3025-200MG
Tris(3,5-dimethyl-1-pyrazolyl)methane ≥98.0% (by GC)
Tris(3,5-dimethyl-1-pyrazolyl)methane
Formula:
C₁₆H₂₂N₆ |
CAS Number:
28791-97-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...