4,4'-Methylenebis(N,N-diglycidylaniline) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4,4-Methylenebis(N,N-bis(oxiran-2-ylmethyl)aniline)
M2619-100G
M2619-25G
TCM2619-100GEA
112.34
CAD
TCM2619-100G
TCM2619-25G
4,4'-Methylenebis(N,N-diglycidylaniline) ≥98.0% (by titrimetric analysis)
4,4'-Methylenebis(N,N-diglycidylaniline)
Formula:
C₂₅H₃₀N₂O₄ MW: 422.53 g/mol Flash Pt: 113°C |
MDL Number:
MFCD00661161 CAS Number: 28768-32-3 UN: 3082 ADR: 9 III |
Specification Test Results
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...