4-Methylumbelliferyl alpha-D-mannopyranoside ≥97.0% (by HPLC)

Supplier: TCI America

M3023-100MG M3023-25MG
TCM3023-100MGEA 466.1 CAD
TCM3023-100MG TCM3023-25MG
4-Methylumbelliferyl alpha-D-mannopyranoside ≥97.0% (by HPLC)
4-Methylumbelliferyl alpha-D-mannopyranoside
Formula: C₁₆H₁₈O₈
Storage Temperature: Freezer
MDL Number: MFCD00067662
CAS Number: 28541-83-5
EINECS: 249-073-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 214.0 to 219.0 deg-C
Specific rotation [a]20/D +157 to +167 deg (C=0.35, H2O)
Specific rotation (value) 157 to 167 deg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR