3-Amino-4-methylphenol ≥98.0% (by GC)

Supplier: TCI America
Warning

A3180-1G A3180-5G
TCA3180-1GEA 96.38 CAD
TCA3180-1G TCA3180-5G
3-Amino-4-methylphenol ≥98.0% (by GC)
3-Amino-4-methylphenol
Formula: C₇H₉NO
MDL Number: MFCD00075083
CAS Number: 2836-00-2
EINECS: 220-622-8

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR