Tris(2,2'-bipyridine)cobalt(III) tris(hexafluorophosphate) ≥93.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3256-1G T3256-200MG
TCT3256-1GEA 1147.08 CAD
TCT3256-1G TCT3256-200MG
Tris(2,2'-bipyridine)cobalt(III) tris(hexafluorophosphate) ≥93.0% (by titrimetric analysis)
Tris(2,2'-bipyridine)cobalt(III) tris(hexafluorophosphate)
Formula: C₃₀H₂₄CoF₁₈N₆P₃
MDL Number: MFCD28976173
CAS Number: 28277-53-4

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Chelometric Titration) min. 93.0 %
Elemental analysis (Nitrogen) 8.10 to 9.30 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR