(2E,4E)-5-Phenyl-2,4-pentadienoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (E,E)-Cinnamylideneacetic acid, (2E,4E)-5-Phenylpenta-2,4-dienoic acid

P2422-25G P2422-5G
76220-428EA 1142.34 CAD
76220-428 76220-430
(2E,4E)-5-Phenyl-2,4-pentadienoic acid ≥98.0% (by GC, titration analysis)
(2E,4E)-5-Phenyl-2,4-pentadienoic acid
Formula: C₁₁H₁₀O₂
MW: 174.20 g/mol
MDL Number: MFCD00014018
CAS Number: 28010-12-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR