trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

P2371-1G P2371-5G
TCP2371-1GEA 52.14 CAD
TCP2371-1G TCP2371-5G
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl
Formula: C₁₈H₃₂
MDL Number: MFCD12761279
CAS Number: 279246-65-0

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR